Learn More
Lalistat 2
Selective LAL inhibitor
Brand: Tocris 6099/10
Additional Details : Weight : 0.00970kg
Description
Lalistat 2 is a selective lysosomal acid lipase inhibitor (IC50 = 152 nM). Exhibits no inhibition of human pancreatic lipase or bovine milk lipoprotein lipase (up to 10 μM).Specifications
4-(Piperidin-1-yl)-1,2,5-thiadiazol-3-yl piperidine-1-carboxylate | |
>98% | |
C13H20N4O2S |
1234569-09-5 | |
10 mg | |
Observed MW: 296.39 |