Learn More
Lalistat 2
Selective LAL inhibitor
Brand:  Tocris 6099/10
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18700328
  • 2846.58 SEK / 10mg
Estimated Shipment: 09-09-2024
to see stock.
Description
Description
Lalistat 2 is a selective lysosomal acid lipase inhibitor (IC50 = 152 nM). Exhibits no inhibition of human pancreatic lipase or bovine milk lipoprotein lipase (up to 10 μM).
Specifications
Specifications
4-(Piperidin-1-yl)-1,2,5-thiadiazol-3-yl piperidine-1-carboxylate
>98%
C13H20N4O2S
1234569-09-5
10 mg
Observed MW: 296.39