missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lalistat 2
Selective LAL inhibitor
2846.58 SEK - 11439.23 SEK
Specifications
Chemical Name or Material | 4-(Piperidin-1-yl)-1,2,5-thiadiazol-3-yl piperidine-1-carboxylate |
---|---|
CAS | 1234569-09-5 |
Assay Percent Range | >98% |
Molecular Formula | C13H20N4O2S |
Formula Weight | Observed MW: 296.39 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18700328
|
Tocris
6099/10 |
10 mg |
2846.58 SEK
10mg |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18700819
|
Tocris
6099/50 |
50 mg |
11439.23 SEK
50mg |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Selective lysosomal acid lipase inhibitor (IC50 = 152 nM). Exhibits no inhibition of human pancreatic lipase or bovine milk lipoprotein lipase (up to 10 μM).Specifications
4-(Piperidin-1-yl)-1,2,5-thiadiazol-3-yl piperidine-1-carboxylate | |
>98% | |
Observed MW: 296.39 |
1234569-09-5 | |
C13H20N4O2S |