Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K27me3 Antibody
The LanthaScreen™ Tb-anti-Histone H3K27me3 antibody is a key component of the BacMam Histone H3K27me3 Cellullar Assay (A14159).
Brand:  Thermo Scientific™ A14152
 View more versions of this product
Product Code. 11652049
  • 351700.00 SEK / 1mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • The LanthaScreen™ Tb-anti-Histone H3K27me3 antibody is a key component of the BacMam Histone H3K27me3 Cellullar Assay (A14159)
  • When this terbium (Tb)- labeled antibody is paired with baculovirus-mediated (BacMam) expression of a Histone H3-GFP fusion protein, the TR-FRET readout enables sensitive detection of H3 lysine 27 trimethylation
  • The resulting high-throughput screening (HTS) compatible assay can be used to identify inhibitors of methyltransferases and demethylases acting on lysine 27 of histone H3
  • For research use only
  • Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K27me3 Antibody Allows Users to:
  • Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
  • Conserve precious cell samples with the miniaturizable homogenous assay format
  • Improve data quality with the advantages of TR-FRET
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3500 nM, 0.5 mg/mL
1 mg
Liquid
Methylated
Tb (Terbium)/Lanthanide Chelates
Store the LanthaScreen™ Tb-anti-histone H3K27me3 Antibody at -20° C.
Dry Ice
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only. Not for use in diagnostic procedures.