missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ Laton Total Nitrogen Cuvette Tests
Brand: Hach Lange™ LCK338
View more versions of this product
This item is not returnable.
View return policy
Specifications
25 | |
15°C to 25°C | |
Laton Total Nitrogen Cuvette Test |
20 to 100mg/L TNb | |
Koroleff Digestion (Peroxodisulphate), and Photometric Detection with 2.6-Dimethylphenol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- LatoN, Sample cuvette
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 3
- Flammable liquid Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H360FD-May damage fertility. May damage the unborn child.
- H226-Flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : Sulphuric acid, Phosphoric acid, Sodiumhydroxide, dipotassium peroxodisulphate,potassium persulpha
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction