Learn More
Hach Lange™ Laton Total Nitrogen Cuvette Tests
2457.00 SEK - 3780.00 SEK
Specifications
No. of Tests | 25 |
---|---|
Storage Requirements | 15°C to 25°C |
Test Method | Koroleff Digestion (Peroxodisulphate), and Photometric Detection with 2.6-Dimethylphenol |
Type | Laton Total Nitrogen Cuvette Test |
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
10329662
|
Hach Lange™
LCK238 |
5 to 40mg/L TNb |
3780.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10409464
|
Hach Lange™
LCK338 |
20 to 100mg/L TNb |
2485.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16411191
|
Hach Lange™
LCK138 |
1 to 16mg/L TNb |
2457.00 SEK
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
25 | |
15°C to 25°C | |
Koroleff Digestion (Peroxodisulphate), and Photometric Detection with 2.6-Dimethylphenol | |
Laton Total Nitrogen Cuvette Test |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- LatoN, Sample cuvette
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 3
- Flammable liquid Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H360FD-May damage fertility. May damage the unborn child.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sulphuric acid, Phosphoric acid, Sodiumhydroxide, Potassiumnitrate, dipotassium peroxodisulphate,