Learn More
Eurofins DiscoverX LATS1 Protein, Active
Active, N-Terminal GST-tagged, recombinant, human LATS1, amino acid residues 589-end and N-Terminal 6His-tagged, recombinant, human MOBKL1A, full length, for use in Kinase Assays.

6046.30 SEK - 46147.75 SEK
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 90 kDa (LATS1) and 30 kDa (MOBKL1A) |
Species | Human |
Recombinant | Recombinant |
Protein Tag | N-terminal GST / N-terminal 6His |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17320884
![]() |
Eurofins DiscoverX
14-988 |
10 μg |
6046.30 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17330884
![]() |
Eurofins DiscoverX
14-988M |
250 μg |
46147.75 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LATS1 is a putative serine/threonine kinase that localizes to the mitotic apparatus and complexes with cell cycle controller CDC2 kinase in early mitosis. The protein is phosphorylated in a cell-cycle dependent manner, with late prophase phosphorylation remaining through metaphase. The N-terminal region of the protein binds CDC2 to form a complex showing reduced H1 histone kinase activity, indicating a role as a negative regulator of CDC2/cyclin A. In addition, the C-terminal kinase domain binds to its own N-terminal region, suggesting potential negative regulation through interference with complex formation via intramolecular binding. Biochemical and genetic data suggest a role as a tumor suppressor. This is supported by studies in knockout mice showing development of soft-tissue sarcomas, ovarian stromal cell tumors and a high sensitivity to carcinogenic treatments.
GenBank NM_004690.2 / GenBank BC038112.1
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
N-terminal GST / N-terminal 6His | |
Truncated | |
LATS1, active |
90 kDa (LATS1) and 30 kDa (MOBKL1A) | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
Combined ≥66% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.