Learn More
Letrazuril, MedChemExpress
MedChemExpress Letrazuril is an anti-HIV agent.
Brand: Medchem Express HY-106859
Description
Letrazuril is an anti-HIV agent.Specifications
Letrazuril | |
Off-White | |
C17H9Cl2FN4O2 | |
DMSO : ≥ 165 mg/mL (421.80 mM) | |
391.18 | |
98.0% | |
Solid |
103337-74-2 | |
Research | |
100 mg | |
O=C(N1)N(N=CC1=O)C2=CC(Cl)=C(C(C3=CC=C(F)C=C3)C#N)C(Cl)=C2 | |
391.18 | |
Research | |
COVID-19-anti-virus |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only