missing translation for 'onlineSavingsMsg'
Learn More

Shamrock™ Litho Color Coded Dot Labels

Offered in a number of colors and diameters to fit your needs. Shamrock™ Color Coded Dot Labels offer multiple opportunities for color coding in the laboratory - test, physician or department.

Brand:  Shamrock™ SDL-100-4

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11706965

  • 184.00 SEK / Pack of 1000
Estimated Shipment: 06-08-2024
to see stock.

Description

Description

  • Resistant to water, oil, and acid
  • Temperature range of -20 to +121°C
  • For autoclave, freezer, refrigerator and oven
  • Universal marking with ballpoint pen, paper pencil and felt
Specifications

Specifications

Permanent
Sheet
Color-coded Dot Label
Red
Litho
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers