Learn More
Shamrock™ Litho Color Coded Dot Labels
Offered in a number of colors and diameters to fit your needs. Shamrock™ Color Coded Dot Labels offer multiple opportunities for color coding in the laboratory - test, physician or department.
125.00 SEK - 184.00 SEK
Specifications
Adhesive Type | Permanent |
---|---|
Material | Litho |
Product Type | Color-coded Dot Label |
Product Code | Brand | Diameter (Metric) | Color | Format | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Color | Format | Price | Quantity & Availability | |||
11776975
|
Shamrock™
SDL-25R-6 |
6.5 mm | Blue | Roll |
138.00 SEK
Pack of 1000 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11772285
|
Shamrock™
SDL-25-6 |
6.5 mm | Blue | Sheet |
138.00 SEK
Pack of 1000 |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11716995
|
Shamrock™
SDL-75R-23 |
19 mm | Brown | Roll |
157.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11786985
|
Shamrock™
SDL-75R-6 |
19 mm | Blue | Roll |
184.00 SEK
Pack of 1000 |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11702295
|
Shamrock™
SDL-25-23 |
6.5 mm | Brown | Sheet |
138.00 SEK
Pack of 1000 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11712295
|
Shamrock™
SDL-25-14 |
6.5 mm | Gray | Sheet |
125.00 SEK
Pack of 1000 |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11746985
|
Shamrock™
SDL-75R-3 |
19 mm | Green | Roll |
166.00 SEK
Pack of 1000 |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11796955
|
Shamrock™
SDL-100-3 |
25 mm | Green | Sheet |
184.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11746975
|
Shamrock™
SDL-25R-3 |
6.5 mm | Green | Roll |
138.00 SEK
Pack of 1000 |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11776985
|
Shamrock™
SDL-75R-5 |
19 mm | Orange | Roll |
184.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11716965
|
Shamrock™
SDL-100-5 |
25 mm | Orange | Sheet |
184.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11762285
|
Shamrock™
SDL-25-5 |
6.5 mm | Orange | Sheet |
138.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11766985
|
Shamrock™
SDL-75R-4 |
19 mm | Red | Roll |
173.00 SEK
Pack of 1000 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11706955
|
Shamrock™
SDL-75-4 |
19 mm | Red | Sheet |
167.00 SEK
Pack of 1000 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11706965
|
Shamrock™
SDL-100-4 |
25 mm | Red | Sheet |
184.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11752285
|
Shamrock™
SDL-25-4 |
6.5 mm | Red | Sheet |
138.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11756975
|
Shamrock™
SDL-25R-4 |
6.5 mm | Red | Roll |
138.00 SEK
Pack of 1000 |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11782285
|
Shamrock™
SDL-25-7 |
6.5 mm | Rose | Sheet |
125.00 SEK
Pack of 1000 |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11796985
|
Shamrock™
SDL-75R-7 |
19 mm | Rose | Roll |
173.00 SEK
Pack of 1000 |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11796935
|
Shamrock™
SDL-50-12 |
13 mm | Violet | Sheet |
125.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11706995
|
Shamrock™
SDL-75R-12 |
19 mm | Violet | Roll |
184.00 SEK
Pack of 1000 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11792285
|
Shamrock™
SDL-25-12 |
6.5 mm | Violet | Sheet |
138.00 SEK
Pack of 1000 |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11726985
|
Shamrock™
SDL-75R-1 |
19 mm | White | Roll |
173.00 SEK
Pack of 1000 |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11776945
|
Shamrock™
SDL-75-1 |
19 mm | White | Sheet |
157.00 SEK
Pack of 1000 |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11776955
|
Shamrock™
SDL-100-1 |
25 mm | White | Sheet |
184.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11726975
|
Shamrock™
SDL-25R-1 |
6.5 mm | White | Roll |
138.00 SEK
Pack of 1000 |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11702285
|
Shamrock™
SDL-25-1 |
6.5 mm | White | Sheet |
125.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11786955
|
Shamrock™
SDL-100-2 |
25 mm | Yellow | Sheet |
184.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11736985
|
Shamrock™
SDL-75R-2 |
19 mm | Yellow | Roll |
157.00 SEK
Pack of 1000 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11712285
|
Shamrock™
SDL-25-2 |
6.5 mm | Yellow | Sheet |
138.00 SEK
Pack of 1000 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11736975
|
Shamrock™
SDL-25R-2 |
6.5 mm | Yellow | Roll |
125.00 SEK
Pack of 1000 |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Resistant to water, oil, and acid
- Temperature range of -20 to +121°C
- For autoclave, freezer, refrigerator and oven
- Universal marking with ballpoint pen, paper pencil and felt
Specifications
Permanent | |
Litho | |
Color-coded Dot Label |