missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Long Ranger™, Long Ranger XL, and Rapid Ranger™ Gel Solutions
A selection of concentrated and premix solutions for automated, high accuracy DNA sequencing applications. Long Ranger™, Long Ranger XL, and Rapid Ranger™ Gel Solutions offer greater flexibility to optimize run conditions and change gel percentages.
Brand: Lonza™ 50611
This item is not returnable.
View return policy
Description
Concentrated Solutions- Offer more flexibility to optimize run conditions and change gel percentages
- Require the addition of urea, buffer, APS, and TEMED
Premix Solutions
- Contain all gel components for increased run-to-run consistency and ease of use
- Require the addition of APS and TEMED for gel polymerization
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Agarose Electrophoresis Gels | |
DNA and RNA Sequencing, genotyping | |
250 mL |
Liquid | |
Agarose | |
18° to 26°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction