missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Long Ranger™, Long Ranger XL, and Rapid Ranger™ Gel Solutions
A selection of concentrated and premix solutions for automated, high accuracy DNA sequencing applications. Long Ranger™, Long Ranger XL, and Rapid Ranger™ Gel Solutions offer greater flexibility to optimize run conditions and change gel percentages.
6475.98 SEK - 21364.92 SEK
Specifications
Product Type | Agarose Electrophoresis Gels |
---|---|
Form | Liquid |
For Use With (Application) | DNA and RNA Sequencing, genotyping |
Material | Agarose |
Storage Requirements | 18° to 26°C |
Description
Concentrated Solutions
- Offer more flexibility to optimize run conditions and change gel percentages
- Require the addition of urea, buffer, APS, and TEMED
Premix Solutions
- Contain all gel components for increased run-to-run consistency and ease of use
- Require the addition of APS and TEMED for gel polymerization
Specifications
Agarose Electrophoresis Gels | |
DNA and RNA Sequencing, genotyping | |
18° to 26°C |
Liquid | |
Agarose |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title