missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ BioWhittaker™ Antibiotics: Gentamicin Sulfate
Used as a broad spectrum bactericidal, with activity against gram negative and gram positive organisms including Pseudomonas. BioWhittaker™ Antibiotics: Gentamicin Sulfate is manufactured in accordance with cGMP regulations and cell culture tested to assure compatibility.
Brand: Lonza™ 17-519Z
This item is not returnable.
View return policy
Description
- Broad spectrum bactericida
- Stable over a wide range of pH and temperature
- Autoclavable
- Colorless to amber or yellow.
- cGMP compatibility
Specifications
15° to 30°C | |
Antibiotic | |
4.2 to 5.7 |
10 mg/mL | |
Screw cap vial | |
10 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Gentamicin Sulfate 10 mg Gentamicin Per ml
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Gentamicin, sulfate(salt)