missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ BioWhittaker™ Antibiotics: Gentamicin Sulfate

Used as a broad spectrum bactericidal, with activity against gram negative and gram positive organisms including Pseudomonas. BioWhittaker™ Antibiotics: Gentamicin Sulfate is manufactured in accordance with cGMP regulations and cell culture tested to assure compatibility.

542.00 SEK - 3697.00 SEK

Specifications

Content And Storage 15° to 30°C
Product Type Antibiotic
View More Specs

Products 5
Product Code Brand Concentration dimForm Quantity Price Quantity & Availability  
Product Code Brand Concentration dimForm Quantity Price Quantity & Availability  
11689980
View Documents Promotion Details
Lonza™
17-518Z
50 mg/mL 10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11679980
View Documents Promotion Details
Lonza™
17-518L
50 mg/mL 10 x 10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11609990
View Documents Promotion Details
Lonza™
17-519Z
10 mg/mL 10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11699980
View Documents Promotion Details
Lonza™
17-519L
10 mg/mL 10 x 10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11619990
View Documents Promotion Details
Lonza™
17-528Z
50 mg/mL 10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Broad spectrum bactericide
  • Stable over a wide range of pH and temperature
  • Autoclavable
  • Colorless to amber or yellow.
  • cGMP compatibility
Specifications

Specifications

15° to 30°C
Antibiotic
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Gentamicin sulfate solution

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : gentamicin sulphate

SDS
Documents

Documents

Product Certifications