Learn More
Lonza™ GelBond™ PAG Film for Acrylamide Gels
Transparent, flexible polyester film supports polyacrylamide or MDE™ gels through electrophoresis, fixing, staining, destaining, and drying procedures
Brand:  Lonza™ 54723
 View more versions of this product
Product Code. 11517765
  • 10245.00 SEK / 50 sheets
Estimated Shipment: 15-11-2024
to see stock.
Explore more special offers
Description
  • Covalently attaches to the acrylamide monomers during the polymerization reaction
  • Makes gels easier to handle
  • Helps gels retain their original dimensions during staining, destaining, and drying
TRUSTED_SUSTAINABILITY
Specifications
Specifications
16 x 18 cm
Polyester
0.007 in.
7 in.
6.3 in.
PAG Film for Acrylamide Gels
50/Pk.
0.02 cm
18 cm
18 cm