Learn More
Lonza™ GelBond™ PAG Film for Acrylamide Gels
Transparent, flexible polyester film supports polyacrylamide or MDE™ gels through electrophoresis, fixing, staining, destaining, and drying procedures
Brand: Lonza™ 54731
This item is not returnable.
View return policy
Description
- Covalently attaches to the acrylamide monomers during the polymerization reaction
- Makes gels easier to handle
- Helps gels retain their original dimensions during staining, destaining, and drying
Specifications
19.9 x 26.4 cm | |
Polyester | |
0.007 in. | |
10.3 in. | |
7.8 in. |
PAG Film for Acrylamide Gels | |
50/Pk. | |
0.02 cm | |
26.4 cm | |
26.4 cm |