Learn More
Fisherbrand™ Magnetic Stir Bar
The ALNICO magnetic stir bar withstands temperatures up to 250°C, making them suitable for use with both unheated and heated stirrers. Smooth shape for better coupling when using concave bottom glassware.
539.00 SEK - 1135.00 SEK
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Coating | PTFE |
Material | Alnico Magnet, PTFE |
Quantity | 10/Cs |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||||
17830752
|
Fisherbrand™
120894 |
5 mm | 10 mm |
539.00 SEK
Pack of 10 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
17820752
|
Fisherbrand™
120895 |
8 mm | 25 mm |
710.00 SEK
Pack of 10 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
17840752
|
Fisherbrand™
120896 |
8 mm | 30 mm |
995.00 SEK
Pack of 10 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
17850752
|
Fisherbrand™
120897 |
8 mm | 35 mm |
1135.00 SEK
Pack of 10 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
Description
- Magnetic bar is made from ALNICO, one of the strongest permanent magnets available
- The sintered manufacturing process reduces the chance of decoupling (spin outs), encapsulation in PTFE protects both magnet and what it is stirring
- Chemically inert and easy to clean
- Can withstand temperatures up to 225°C, making them suitable for use with both unheated and heated stirrers
Specifications
Not Autoclavable | |
PTFE | |
10/Cs | |
Stir Bar | |
Stirrers |
White | |
Alnico Magnet, PTFE | |
Smooth | |
Stirring solutions | |
Up to 250°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Magnetic Stir Bar