Learn More
Magnolin, MedChemExpress
MedChemExpress Magnolin, a major component of Magnolia flos (Shin-Yi), inhibits the Ras/ERKs/RSK2 signaling axis by targeting the active pocket of ERK1 and ERK2 with IC50s of 87 nM and 16.5 nM, respectively.
Brand: Medchem Express HY-N1374
This item is not returnable.
View return policy
Description
Magnolin, a major component of Magnolia flos (Shin-Yi), inhibits the Ras/ERKs/RSK2 signaling axis by targeting the active pocket of ERK1 and ERK2 with IC50s of 87 nM and 16.5 nM, respectively.Specifications
Magnolin | |
White | |
C23H28O7 | |
DMSO : 100 mg/mL (240.12 mM; Need ultrasonic) | |
416.46 | |
98.85% | |
Solid |
31008-18-1 | |
Research | |
10 mg | |
COC1=C(OC)C(OC)=CC([C@H]2OC[C@]3([H])[C@@H](C4=CC=C(OC)C(OC)=C4)OC[C@@]32[H])=C1 | |
416.46 | |
Research | |
COVID-19-immunoregulation |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.