Learn More
Magnolin, MedChemExpress
MedChemExpress Magnolin, a major component of Magnolia flos (Shin-Yi), inhibits the Ras/ERKs/RSK2 signaling axis by targeting the active pocket of ERK1 and ERK2 with IC50s of 87 nM and 16.5 nM, respectively.
POA - 2056.31 SEK
Specifications
Chemical Name or Material | Magnolin |
---|---|
CAS | 31008-18-1 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C23H28O7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16463916
|
Medchem Express
HY-N1374 |
25 mg |
2056.31 SEK
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16451628
|
Medchem Express
HY-N1374 |
5 mg |
581.13 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16419218
|
Medchem Express
HY-N1374 |
10 mg |
1028.15 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16404917
|
Medchem Express
HY-N1374 |
50 mg |
POA
50mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Magnolin, a major component of Magnolia flos (Shin-Yi), inhibits the Ras/ERKs/RSK2 signaling axis by targeting the active pocket of ERK1 and ERK2 with IC50s of 87 nM and 16.5 nM, respectively.Specifications
Magnolin | |
White | |
C23H28O7 | |
COC1=C(OC)C(OC)=CC([C@H]2OC[C@]3([H])[C@@H](C4=CC=C(OC)C(OC)=C4)OC[C@@]32[H])=C1 | |
416.46 | |
Research | |
COVID-19-immunoregulation |
31008-18-1 | |
Research | |
DMSO : 100 mg/mL (240.12 mM; Need ultrasonic) | |
416.46 | |
98.85% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.