Learn More
Magnolin, MedChemExpress
MedChemExpress Magnolin, a major component of Magnolia flos (Shin-Yi), inhibits the Ras/ERKs/RSK2 signaling axis by targeting the active pocket of ERK1 and ERK2 with IC50s of 87 nM and 16.5 nM, respectively.
Brand:  Medchem Express HY-N1374
Encompass_Preferred
 View more versions of this product
Product Code. 16451628
  • 581.13 SEK / 5mg
Explore more special offers
Description
Description
Magnolin, a major component of Magnolia flos (Shin-Yi), inhibits the Ras/ERKs/RSK2 signaling axis by targeting the active pocket of ERK1 and ERK2 with IC50s of 87 nM and 16.5 nM, respectively.
Specifications
Specifications
Magnolin
White
C23H28O7
DMSO : 100 mg/mL (240.12 mM; Need ultrasonic)
416.46
98.85%
Solid
31008-18-1
Research
5 mg
COC1=C(OC)C(OC)=CC([C@H]2OC[C@]3([H])[C@@H](C4=CC=C(OC)C(OC)=C4)OC[C@@]32[H])=C1
416.46
Research
COVID-19-immunoregulation
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.