missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAST1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13305-1-AP-150UL
This item is not returnable.
View return policy
Description
Appears to link the dystrophin/utrophin network with microtubule filaments via the syntrophins. Phosphorylation of DMD or UTRN may modulate their affinities for associated proteins.Specifications
MAST1 | |
Polyclonal | |
Unconjugated | |
MAST1 | |
KIAA0973, MAST1, SAST, SAST170 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22983, 353118, 56527 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q810W7, Q9R1L5, Q9Y2H9 | |
MAST1 | |
MAST1 Fusion Protein Ag4126 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |