missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAST1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | MAST1 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881444
|
Proteintech
13305-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16871444
|
Proteintech
13305-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Appears to link the dystrophin/utrophin network with microtubule filaments via the syntrophins. Phosphorylation of DMD or UTRN may modulate their affinities for associated proteins.Specifications
MAST1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q810W7, Q9R1L5, Q9Y2H9 | |
22983, 353118, 56527 | |
MAST1 Fusion Protein Ag4126 | |
Primary | |
-20°C | |
MAST1 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0973, MAST1, SAST, SAST170 | |
MAST1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |