Learn More
Maxacalcitol-d6, MedChemExpress
MedChemExpress Maxacalcitol-D6 is the deuterated form of Maxacalcitol (22-Oxacalcitriol), which is a non-calcemic vitamin D3 analog and VDR ligand of VDR-like receptors.
Brand:  Medchem Express HY-15329
Encompass_Preferred
 View more versions of this product
Product Code. 16411509
  • 29026.65 SEK / 1mg
Explore more special offers
Description
Description
Maxacalcitol-D6 is the deuterated form of Maxacalcitol (22-Oxacalcitriol), which is a non-calcemic vitamin D3 analog and VDR ligand of VDR-like receptors.
Specifications
Specifications
Maxacalcitol-d6
White
C26H36D6O4
424.65
96.8%
Solid
1 mg
Research
O[C@@H](C[C@H](O)C/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](OCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2
424.65
Research