missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maxacalcitol-d6, MedChemExpress
MedChemExpress Maxacalcitol-D6 is the deuterated form of Maxacalcitol (22-Oxacalcitriol), which is a non-calcemic vitamin D3 analog and VDR ligand of VDR-like receptors.
29026.65 SEK - 96765.41 SEK
Specifications
Molecular Formula | C26H36D6O4 |
---|---|
Molecular Weight (g/mol) | 424.65 |
Percent Purity | 96.8% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16449047
|
Medchem Express
HY-15329 |
5 mg |
96765.41 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16411509
|
Medchem Express
HY-15329 |
1 mg |
29026.65 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Maxacalcitol-D6 is the deuterated form of Maxacalcitol (22-Oxacalcitriol), which is a non-calcemic vitamin D3 analog and VDR ligand of VDR-like receptors.Specifications
C26H36D6O4 | |
424.65 | |
96.8% |
Safety and Handling
missing translation for 'shelfLife' : 4°C, protect from light, stored under nitrogen∣The compound is unstable in solutions, freshly prepared is recommended.
Recommended Storage : 4°C, protect from light, stored under nitrogen∣The compound is unstable in solutions, freshly prepared is recommended.
Research purposes only