Learn More
Maxacalcitol-d6, MedChemExpress
MedChemExpress Maxacalcitol-D6 is the deuterated form of Maxacalcitol (22-Oxacalcitriol), which is a non-calcemic vitamin D3 analog and VDR ligand of VDR-like receptors.
Brand: Medchem Express HY-15329
This item is not returnable.
View return policy
Description
Maxacalcitol-D6 is the deuterated form of Maxacalcitol (22-Oxacalcitriol), which is a non-calcemic vitamin D3 analog and VDR ligand of VDR-like receptors.Specifications
Maxacalcitol-d6 | |
White | |
C26H36D6O4 | |
424.65 | |
96.8% | |
Solid |
5 mg | |
Research | |
O[C@@H](C[C@H](O)C/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](OCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2 | |
424.65 | |
Research |
Research purposes only