Learn More
Mericitabine, MedChemExpress
MedChemExpress Mericitabine (RG 7128; R-7128) is a nucleoside inhibitor of the HCV NS5B polymerase that acts as an RNA chain terminator and prevents elongation of RNA transcripts during replication.
Brand:  Medchem Express HY-10240
Encompass_Preferred
 View more versions of this product
Product Code. 16403738
  • 4544.72 SEK / 10mg
Explore more special offers
Description
Description
Mericitabine (RG 7128
Specifications
Specifications
Mericitabine
White
C18H26FN3O6
RG 7128 R-7128 PSI 6130 diisobutyrate
O=C1N=C(C=CN1[C@@H]([C@@](C)([C@@H]2OC(C(C)C)=O)F)O[C@@H]2COC(C(C)C)=O)N
399.41
Research
COVID-19-anti-virus
940908-79-2
Research
10 mg
DMSO : 100 mg/mL (250.37 mM; Need ultrasonic)
399.41
99.47%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.