Learn More
Mericitabine, MedChemExpress
MedChemExpress Mericitabine (RG 7128; R-7128) is a nucleoside inhibitor of the HCV NS5B polymerase that acts as an RNA chain terminator and prevents elongation of RNA transcripts during replication.
3293.07 SEK - 11726.88 SEK
Specifications
Chemical Name or Material | Mericitabine |
---|---|
CAS | 940908-79-2 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C18H26FN3O6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16460117
|
Medchem Express
HY-10240 |
50 mg |
11726.88 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16426467
|
Medchem Express
HY-10240 |
5 mg |
3293.07 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16403738
|
Medchem Express
HY-10240 |
10 mg |
4544.72 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mericitabine (RG 7128Specifications
Mericitabine | |
White | |
C18H26FN3O6 | |
DMSO : 100 mg/mL (250.37 mM; Need ultrasonic) | |
399.41 | |
99.47% | |
Solid |
940908-79-2 | |
Research | |
RG 7128 R-7128 PSI 6130 diisobutyrate | |
O=C1N=C(C=CN1[C@@H]([C@@](C)([C@@H]2OC(C(C)C)=O)F)O[C@@H]2COC(C(C)C)=O)N | |
399.41 | |
Research | |
COVID-19-anti-virus |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.