Learn More
Mericitabine, MedChemExpress
MedChemExpress Mericitabine (RG 7128; R-7128) is a nucleoside inhibitor of the HCV NS5B polymerase that acts as an RNA chain terminator and prevents elongation of RNA transcripts during replication.
Brand: Medchem Express HY-10240
This item is not returnable.
View return policy
Description
Mericitabine (RG 7128Specifications
Mericitabine | |
White | |
C18H26FN3O6 | |
RG 7128 R-7128 PSI 6130 diisobutyrate | |
O=C1N=C(C=CN1[C@@H]([C@@](C)([C@@H]2OC(C(C)C)=O)F)O[C@@H]2COC(C(C)C)=O)N | |
399.41 | |
Research | |
COVID-19-anti-virus |
940908-79-2 | |
Research | |
10 mg | |
DMSO : 100 mg/mL (250.37 mM; Need ultrasonic) | |
399.41 | |
99.47% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.