missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 1-methylcyclopropyl ketone, 95%, Thermo Scientific Chemicals
1001.14 SEK - 8841.00 SEK
Chemical Identifiers
CAS | 1567-75-5 |
---|---|
Molecular Formula | C6H10O |
Molecular Weight (g/mol) | 98.14 |
InChI Key | OQBCJXUAQQMTRW-UHFFFAOYSA-N |
Synonym | methyl 1-methylcyclopropyl ketone, 1-1-methylcyclopropyl ethan-1-one, 1-1-methylcyclopropyl ethanone, 1-acetyl-1-methylcyclopropane, ethanone, 1-1-methylcyclopropyl, 1-methylcyclopropyl methyl ketone, methyl 1-methyl cyclopropyl ketone, 1-1-methyl-cyclopropyl-ethanone, methyl1-methylcyclopropylketone |
PubChem CID | 74067 |
IUPAC Name | 1-(1-methylcyclopropyl)ethanone |
SMILES | CC(=O)C1(CC1)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10103580
|
Thermo Scientific Acros
147630050 |
5 g | Glass bottle |
1001.14 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10307030
|
Thermo Scientific Acros
147630250 |
25 g | Glass bottle |
4023.48 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904841
|
Thermo Scientific Acros
147631000 |
100 g | Glass Bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1567-75-5 | |
98.14 | |
methyl 1-methylcyclopropyl ketone, 1-1-methylcyclopropyl ethan-1-one, 1-1-methylcyclopropyl ethanone, 1-acetyl-1-methylcyclopropane, ethanone, 1-1-methylcyclopropyl, 1-methylcyclopropyl methyl ketone, methyl 1-methyl cyclopropyl ketone, 1-1-methyl-cyclopropyl-ethanone, methyl1-methylcyclopropylketone | |
1-(1-methylcyclopropyl)ethanone |
C6H10O | |
OQBCJXUAQQMTRW-UHFFFAOYSA-N | |
74067 | |
CC(=O)C1(CC1)C |
Specifications
1567-75-5 | |
0.8950g/mL | |
24°C | |
94% min. (GC) | |
CH3C3H4COCH3 | |
0.895 | |
Solubility in water: insoluble. | |
CC(=O)C1(CC1)C | |
98.14 | |
98.14 | |
Liquid |
Colorless to Yellow | |
125.0°C to 128.0°C | |
Authentic | |
C6H10O | |
1.4330 to 1.4350 | |
methyl 1-methylcyclopropyl ketone, 1-1-methylcyclopropyl ethan-1-one, 1-1-methylcyclopropyl ethanone, 1-acetyl-1-methylcyclopropane, ethanone, 1-1-methylcyclopropyl, 1-methylcyclopropyl methyl ketone, methyl 1-methyl cyclopropyl ketone, 1-1-methyl-cyclopropyl-ethanone, methyl1-methylcyclopropylketone | |
OQBCJXUAQQMTRW-UHFFFAOYSA-N | |
1-(1-methylcyclopropyl)ethanone | |
74067 | |
95% | |
Methyl 1-methylcyclopropyl ketone |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 216-369-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methyl 1-methylcyclopropyl ketone
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only