missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-cyclopentenecarboxylate, 97%, Thermo Scientific Chemicals

490.00 SEK - 1283.25 SEK

Chemical Identifiers

CAS 58101-60-3
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD04038661
InChI Key CEOILRYKIJRPBZ-UHFFFAOYSA-N
Synonym methyl 3-cyclopentenecarboxylate, methyl cyclopent-3-enecarboxylate, methyl-3-cyclopentene-1-carboxylate, methyl3-cyclopentenecarboxylate, 3-cyclopentene-1-carboxylic acid methyl ester, 3-cyclopentene-1-carboxylic acid, methyl ester, acmc-209m2j, methyl cyclopent-3-ene carboxylate, 4-methoxycarbonyl cyclopent-1-ene, methyl 3-cyclopentene-1-carboxylate
PubChem CID 1514118
IUPAC Name methyl cyclopent-3-ene-1-carboxylate
SMILES COC(=O)C1CC=CC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15565966
View Documents
Thermo Scientific Acros
457400010
1 g N/A N/A N/A
15575966
View Documents
Thermo Scientific Acros
457400050
5 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 3-cyclopentenecarboxylate, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.