missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Rabbit anti-Human, Clone: 3I1C1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16696-100UL
This item is not returnable.
View return policy
Description
MHC Class I Monoclonal antibody specifically detects MHC Class I in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
MHC Class I | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
3I1C1 | |
Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human MHC Class I (P04439). KETLQRTDPPKTHMTHHPISDHEATLRCWALGFYPAEITLTWQRDGEDQTQDTELVETRPAGDGTFQKWAAVVVPSGEEQRYTCHVQHEGLPKPLTLRWEL | |
100 μg | |
Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Stem Cell Markers | |
3133 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction