missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I Rabbit anti-Human, Clone: 3I1C1, Novus Biologicals™
Rabbit Monoclonal Antibody
1950.00 SEK - 4805.00 SEK
Specifications
Antigen | MHC Class I |
---|---|
Clone | 3I1C1 |
Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18332125
|
Bio-Techne
NBP3-16696-20UL |
20 μg |
1950.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18306395
|
Bio-Techne
NBP3-16696-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MHC Class I Monoclonal antibody specifically detects MHC Class I in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
MHC Class I | |
Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human MHC Class I (P04439). KETLQRTDPPKTHMTHHPISDHEATLRCWALGFYPAEITLTWQRDGEDQTQDTELVETRPAGDGTFQKWAAVVVPSGEEQRYTCHVQHEGLPKPLTLRWEL | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
3I1C1 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3133 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title