missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ MiniMark™ High Performance Ribbons
MiniMark™ High Performance Ribbons for printing on B-595 Vinyl, excellent solvent resistance, high heat resistance, and super scratch resistance.
Brand: Brady™ 710346
Additional Details : Weight : 0.10100kg
Description
- Excellent solvent resistance, high heat resistance, and super scratch resistance.
- High Performance Resin Ribbon for printing on B-595 Vinyl.
- 110 mm Wide
Specifications
Printer Ribbon | |
MiniMark Printer | |
4.33 in. | |
110 mm | |
90 m x 110 mm | |
Ink side out |
Green | |
98.4 yd. | |
90 m | |
12.7 mm I.D. | |
110 mm Green Ribbon | |
1 Ea. |