Learn More
MK-5046, MedChemExpress
MedChemExpress MK-5046 is a novel BRS-3 agonist, binds to BRS-3 with high affinity (mouse Ki = 1.6 nM, human Ki = 25 nM).
Brand: Medchem Express HY-14342
This item is not returnable.
View return policy
Description
MK-5046 is a novel BRS-3 agonist, binds to BRS-3 with high affinity (mouse Ki = 1.6 nM, human Ki = 25 nM).Specifications
MK-5046 | |
1022152-70-0 | |
5 mg | |
C20H18F6N4O | |
O[C@](CC1=NC(CC2(C(F)(F)F)CC2)=CN1)(C(F)(F)F)C3=CC=C(N4C=CC=N4)C=C3 | |
444.37 | |
Research |
Powder | |
Light Yellow | |
Research | |
DMSO : 14.29 mg/mL (32.16 mM; Need ultrasonic) | |
444.37 | |
99.47% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MK-5046, MedChemExpress