Learn More
MK-5046, MedChemExpress
MedChemExpress MK-5046 is a novel BRS-3 agonist, binds to BRS-3 with high affinity (mouse Ki = 1.6 nM, human Ki = 25 nM).
Brand:  Medchem Express HY-14342
Encompass_Preferred
 View more versions of this product
Product Code. 16418647
  • 3532.26 SEK / 5mg
Explore more special offers
Description
Description
MK-5046 is a novel BRS-3 agonist, binds to BRS-3 with high affinity (mouse Ki = 1.6 nM, human Ki = 25 nM).
Specifications
Specifications
MK-5046
1022152-70-0
5 mg
C20H18F6N4O
O[C@](CC1=NC(CC2(C(F)(F)F)CC2)=CN1)(C(F)(F)F)C3=CC=C(N4C=CC=N4)C=C3
444.37
Research
Powder
Light Yellow
Research
DMSO : 14.29 mg/mL (32.16 mM; Need ultrasonic)
444.37
99.47%
Documents
One moment while we fetch your results.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MK-5046, MedChemExpress