Learn More
MK-5108, MedChemExpress
MedChemExpress MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.
Brand:  Medchem Express HY-13252
Encompass_Preferred
 View more versions of this product
Product Code. 16431757
  • 7235.68 SEK / 50mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.
Specifications
Specifications
MK-5108
White
C22H21ClFN3O3S
VX-689
FC(C(Cl)=CC=C1)=C1O[C@H](CC2)CC[C@@]2(C(O)=O)CC3=CC=CC(NC4=NC=CS4)=N3
461.94
Research
Cancer-Kinase/protease
1010085-13-8
Research
50 mg
DMSO : 12.5 mg/mL (27.06 mM; Need ultrasonic)
461.94
96.69%
Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MK-5108, MedChemExpress