missing translation for 'onlineSavingsMsg'
Learn More

MK-5108, MedChemExpress

MedChemExpress MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.

Encompass_Preferred

1602.75 SEK - 12059.47 SEK

Specifications

Chemical Name or Material MK-5108
CAS 1010085-13-8
Color White
Purity Grade Notes Research
Molecular Formula C22H21ClFN3O3S
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16465588
Encompass_Preferred
View Documents
Medchem Express
HY-13252
10 mg
2738.66 SEK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16438287
Encompass_Preferred
View Documents
Medchem Express
HY-13252
5 mg
1602.75 SEK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16431757
Encompass_Preferred
View Documents
Medchem Express
HY-13252
50 mg
7235.68 SEK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16406136
Encompass_Preferred
View Documents
Medchem Express
HY-13252
100 mg
12059.47 SEK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.
Specifications

Specifications

MK-5108
White
C22H21ClFN3O3S
DMSO : 12.5 mg/mL (27.06 mM; Need ultrasonic)
461.94
96.69%
Powder
1010085-13-8
Research
VX-689
FC(C(Cl)=CC=C1)=C1O[C@H](CC2)CC[C@@]2(C(O)=O)CC3=CC=CC(NC4=NC=CS4)=N3
461.94
Research
Cancer-Kinase/protease
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
MK-5108, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.