Learn More
Köttermann Mobile Cabinet with Two Doors
This Mobile Cabinet with Two Doors offers versatile storage space with two doors to protect stored items.
Brand: Köttermann 315-00140
8731.65 SEK valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Versatile storage space
- Mobile with lockable wheels
- Features two doors to protect stored items
- Many sizes and configurations available
Specifications
White | |
516 x 900 x 740 mm | |
Epoxy Coated | |
Left and Right | |
2 | |
None | |
Yes |
Mobile | |
Hinged | |
Steel | |
2 | |
None | |
Plastic with Brakes | |
Steel |