Learn More
MOBS, 0.2M buffer soln., pH 8.5, Thermo Scientific™
673.34 SEK - 698.00 SEK
Specifications
CAS | 115724-21-5 |
---|---|
Color | Colorless |
Concentration | 0.2 M |
pH | 8.5 |
Physical Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15424619
|
Thermo Scientific Alfa Aesar
J62203.AE |
100 mL |
673.34 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15434619
|
Thermo Scientific Alfa Aesar
J62203.AK |
250 mL | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
115724-21-5 | |
0.2 M | |
Liquid | |
MFCD00674916 | |
VTOWJTPBPWTSMK-UHFFFAOYSA-N | |
4-(morpholin-4-yl)butane-1-sulfonic acid |
Colorless | |
8.5 | |
C8H17NO4S | |
4-(4-Morpholinyl)butanesulfonic acid | |
OS(=O)(=O)CCCCN1CCOCC1 | |
223.29 |
Safety and Handling
Recommended Storage : Keep cold