missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse BD-3, Animal-Free Recombinant Protein, PeproTech®
Recombinant Protein
Brand: Gibco™ AF-250-41-250UG
This item is not returnable.
View return policy
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].Specifications
Functional Assay | |
protein with no preservative | |
HPLC, SDS-PAGE | |
-20°C | |
<0.1 EU/ μg | |
Protein | |
Unconjugated | |
KKINNPVSCL RKGGRCWNRC IGNTRQIGSC GVPFLKCCKR K | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Lyophilized | |
4.6 kDa | |
250 μg | |
RUO | |
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103 A; defensin, beta 103; defensin, beta 103 A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
Mouse | |
Recombinant | |
E. coli |