missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse BD-3, Animal-Free Recombinant Protein, PeproTech®
Recombinant Protein
1020.00 SEK - 11826.00 SEK
Specifications
For Use With (Application) | Functional Assay |
---|---|
Format | Lyophilized |
Formulation | protein with no preservative |
Molecular Weight (g/mol) | 4.6 kDa |
Purification Method | HPLC, SDS-PAGE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17836638
|
Gibco™
AF-250-41-250UG |
250 μg |
11826.00 SEK
250µg |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17897058
|
Gibco™
AF-250-41-50UG |
50 μg |
4209.00 SEK
50µg |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17886578
|
Gibco™
AF-250-41-5UG |
5 μg |
1020.00 SEK
5µg |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].Specifications
Functional Assay | |
protein with no preservative | |
HPLC, SDS-PAGE | |
RUO | |
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103 A; defensin, beta 103; defensin, beta 103 A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
Mouse | |
Recombinant | |
E. coli |
Lyophilized | |
4.6 kDa | |
-20°C | |
<0.1 EU/ μg | |
Protein | |
Unconjugated | |
KKINNPVSCL RKGGRCWNRC IGNTRQIGSC GVPFLKCCKR K | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |