Learn More
MP Biomedicals™ Casein Hydrolysate Enzymatic, Soluble, Powder
Soluble; NZ Amine A ; Peptone from casein
Brand: MP Biomedicals™ 0296013801
Additional Details : Weight : 0.01000kg
Description
Casein hydrolysate enzymatic is recommended for use as an animal diet.
Specifications
Animal Diet | |
Powder | |
454 g |
Moisture 3.5%, Amino N/Total N 50.3%, Chloride 0.8%, Iron <50 ppm, Solubility 25 g/L, Total Nitro gen 12.8%, Sodium 2.8%, Calcium 0.1%, pH (2% solution) 6.6, Ammonia 0.9 | |
Casein Hydrolysate Enzymatic is recommended for use as animal diet, also recommended for the production of media for the growth of Charon phage and other lambda phages | |
15°C to 30°C |