Learn More
MP Biomedicals™ Casein Hydrolysate Enzymatic, Soluble, Powder
Soluble; NZ Amine A ; Peptone from casein
Brand:  MP Biomedicals™ 0296013805
 View more versions of this product
Product Code. 11412841
  • 11440.00 SEK / 5lb
Explore more special offers
This item is not returnable. View return policy
Description
Description
Casein hydrolysate enzymatic is recommended for use as an animal diet.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Animal Diet
Powder
5 lb.
Moisture 3.5%, Amino N/Total N 50.3%, Chloride 0.8%, Iron <50 ppm, Solubility 25 g/L, Total Nitro gen 12.8%, Sodium 2.8%, Calcium 0.1%, pH (2% solution) 6.6, Ammonia 0.9
Casein Hydrolysate Enzymatic is recommended for use as animal diet, also recommended for the production of media for the growth of Charon phage and other lambda phages
15°C to 30°C