Learn More
MP Biomedicals™ Elastase 2X Crystallized, from Porcine Pancreas
Activity 50 u/mg protein, Porcine Pancreas, Milky Solution
Brand: MP Biomedicals™ 0210061710
Description
Elastase was used to study human emphysema by induction of emphysema in rodents. It has been used in a study to investigate the design, synthesis and evaluation of biomimetic affinity ligands for elastases. Elastase from porcine pancreas has also been used in a study to investigate the purification and partial characterization of the pancreatic proteolytic enzymes trypsin, chymotrypsin, and elastase. It hydrolyzes peptide bonds, especially those adjacent to neutral amino acids.

Specifications
2X | |
Investigate the purification and partial characterization of the pancreatic proteolytic Enzymes | |
Porcine pancreas | |
Elastase | |
DFP, Elastinal, alpha-2-macroglobulin |
Milky | |
4°C | |
10 mg | |
≈25000 | |
Solution |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.