Learn More
MP Biomedicals™ Mouse Serum
Used for blocking agents and controls in immunoassays.
509.86 SEK - 1404.50 SEK
Product Code | Brand | Form | Hemoglobin Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Hemoglobin Concentration | Quantity | Price | Quantity & Availability | |||
11408541
|
MP Biomedicals™
0215228201 |
Liquid | 9.65 mg/dL | 1 mL |
509.86 SEK
1mL |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||
11418541
|
MP Biomedicals™
0215228205 |
Liquid | 9.65 mg/dL | 5 mL |
1150.10 SEK
5mL |
Estimated Shipment: 31-12-2024
Log in to see stock. |
|||
11423976
|
MP Biomedicals™
08642931 |
Lyophilized | 5.8 g/dL | 10 mL |
1404.50 SEK
10mL |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||