Learn More
Hanna Instruments™ Multiparameter Chemical Test Kit
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Multiparameter Chemical Test Kit features pre-made, pre-measured titration reagents for the determination of multiple parameters commonly measured for environmental/facility monitoring.
Brand: Hanna Instruments™ HI3821
Description
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
- Complete setup: All required materials are included, such as beakers, syringes, a case and pre-measured reagent bottles and packets
- High resolution readings for fast, accurate determinations
- Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
Cooling and Boiler Monitoring | |
All necessary reagents and accessories for each test, DO glass bottle, sample beaker, color comparison cube, hard carrying case, and instructions | |
1 Kit | |
Testing of: alkalinity, chloride, hardness, dissolved oxygen, phosphate, and sulfite | |
Chemical Test Kit |
Water samples | |
5 parameters x 100 tests each, 1 parameter (phosphate) x 50 tests | |
Various | |
Titration and colorimetry |
Safety and Handling
- COOLING AND BOILER WATER TEST KIT
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H300-Fatal if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H332-Harmful if inhaled.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- EUH032-Contact with acids liberates very toxic gas.
- EUH071-Corrosive to the respiratory tract.
- EUH208-Contains . May produce an allergic reaction.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use powder to extinguish.
- P390-Absorb spillage to prevent material damage.
- P391-Collect spillage.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydroxide, Potassium iodide, Sodium azide, Propionicacid, Ethanol, Hydrochloric acid, Mercur
Your input is important to us. Please complete this form to provide feedback related to the content on this product.