Learn More
n-Dodecylpyridinium chloride hydrate, 98%, Thermo Scientific Chemicals
820.00 SEK - 1730.00 SEK
Chemical Identifiers
CAS | 207234-02-4 |
---|---|
Molecular Formula | C17H30ClN |
Molecular Weight (g/mol) | 283.88 |
MDL Number | MFCD00011987 |
InChI Key | GKQHIYSTBXDYNQ-UHFFFAOYSA-M |
Synonym | 1-dodecylpyridin-1-ium chloride hydrate, laurylpyridinum chloride, 1-dodecylpyridinium chloride hydrate, 1-dodecylpyridin-1-ium hydrate chloride, c17h30n.cl.h2o, n-dodecylpyridinium chloride hydrate, dodecylpyridinium chloride monohydrate, n-dodecylpyridinium chloride monohydrate, 1-dodecylpyridin-1-ium chloride-water 1/1/1 |
PubChem CID | 16211802 |
IUPAC Name | 1-dodecylpyridin-1-ium;chloride;hydrate |
SMILES | [Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435497
|
Thermo Scientific Alfa Aesar
B23492.14 |
25 g |
820.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445497
|
Thermo Scientific Alfa Aesar
B23492.22 |
100 g |
1730.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
207234-02-4 | |
283.88 | |
GKQHIYSTBXDYNQ-UHFFFAOYSA-M | |
16211802 | |
[Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1 |
C17H30ClN | |
MFCD00011987 | |
1-dodecylpyridin-1-ium chloride hydrate, laurylpyridinum chloride, 1-dodecylpyridinium chloride hydrate, 1-dodecylpyridin-1-ium hydrate chloride, c17h30n.cl.h2o, n-dodecylpyridinium chloride hydrate, dodecylpyridinium chloride monohydrate, n-dodecylpyridinium chloride monohydrate, 1-dodecylpyridin-1-ium chloride-water 1/1/1 | |
1-dodecylpyridin-1-ium;chloride;hydrate |
Specifications
207234-02-4 | |
C17H30ClN | |
Hygroscopic | |
GKQHIYSTBXDYNQ-UHFFFAOYSA-M | |
1-dodecylpyridin-1-ium;chloride;hydrate | |
16211802 | |
98% |
88°C to 90°C | |
MFCD00011987 | |
1-dodecylpyridin-1-ium chloride hydrate, laurylpyridinum chloride, 1-dodecylpyridinium chloride hydrate, 1-dodecylpyridin-1-ium hydrate chloride, c17h30n.cl.h2o, n-dodecylpyridinium chloride hydrate, dodecylpyridinium chloride monohydrate, n-dodecylpyridinium chloride monohydrate, 1-dodecylpyridin-1-ium chloride-water 1/1/1 | |
[Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1 | |
283.88 | |
283.89 (Anhydrous) | |
n-Dodecylpyridinium chloride hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 203-232-2
missing translation for 'rtecsNumber' : UU4017070
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.