Learn More
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
Brand: Thermo Scientific™ C4010-17AW
This item is not returnable.
View return policy
Description
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
- Assembled kits include 100 vials with pre-attached caps and septa
- Packaged in convenient vial trays with clear covers or in polybags
- Vial trays are stackable for easy storage
- Vial trays hold vials upright for easy filling and maintaining sample order
Specifications
Light Blue | |
Amber | |
Red PTFE/White Silicone |
Amber Glass I-D; Light Blue Cap; Red PTFE/White Silicone septum | |
PTFE, Silicone | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits > Amber Glass I-D; Light Blue Cap; Red PTFE/White Silicone septum
Spot an opportunity for improvement?Share a Content Correction