Learn More
Product Code | Brand | Description | Closure Color | Septum | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Closure Color | Septum | Color | Price | Quantity & Availability | |||||
12463608
|
Thermo Scientific™
C4010-018 |
Clear Glass; Light Blue Cap; PTFE/Silicone Pre-slit septum | Light Blue | PTFE/Silicone | Clear |
769.40 SEK
Pack of 100 |
Estimated Shipment: 04-04-2025
Log in to see stock. |
|||||
12433628
|
Thermo Scientific™
C4010-17A |
Amber Glass; Light Blue Cap; Red PTFE/White Silicone septum | Light Blue | Red PTFE/White Silicone | Amber | N/A | ||||||
12453628
|
Thermo Scientific™
C4010-17AW |
Amber Glass I-D; Light Blue Cap; Red PTFE/White Silicone septum | Light Blue | Red PTFE/White Silicone | Amber | N/A | ||||||
12463628
|
Thermo Scientific™
C4010-17BLK |
Clear Glass; Black cap; PTFE/Silicone septum | Light Blue | Red PTFE/White Silicone | Clear | N/A | ||||||
Description
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
- Assembled kits include 100 vials with pre-attached caps and septa
- Packaged in convenient vial trays with clear covers or in polybags
- Vial trays are stackable for easy storage
- Vial trays hold vials upright for easy filling and maintaining sample order
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits