missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ I-Series Stackable Shakers: Stacking Kits
For use only with I-Series Stackable Shakers, Models I-26 and I-26R
Brand: Eppendorf™ M1324-0800
Requires : Sticky Pad Platform (14-285-746) requires Sticky Pad (14-285-753, sold separately)
This item is not returnable. View return policy
Description
- Stacking kit enables two I-26 Series Stackable Shakers to be stacked vertically
- Bases allow shaker(s) to be raised to suitable height
Specifications
125 mL to 2.7 L | |
13 in. | |
NBS I-Series Stackable Shakers |
Base Riser for two stack | |
33.02 cm | |
Medium |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction