Learn More
Eppendorf™ I-Series Stackable Shakers: Stacking Kits
For use only with I-Series Stackable Shakers, Models I-26 and I-26R
Brand:  Eppendorf™ M1324-0500
 View more versions of this product
Product Code. 11682754
  • 5085.26 SEK / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Requires : Sticky Pad Platform (14-285-746) requires Sticky Pad (14-285-753, sold separately)
This item is not returnable. View return policy
Description
Description
  • Stacking kit enables two I-26 Series Stackable Shakers to be stacked vertically
  • Bases allow shaker(s) to be raised to suitable height
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stacking Kit (to join two shakers)
NBS I-Series Stackable Shakers